Learn More
Emma Safety Footwear Vila Safety Shoes
Lace-up shoe with zipped cover provides additional protection against all kinds of dirt, germs and bacteria. Features SmartecAir™ for dry, fresh feet.
Brand:  Emma Safety Footwear MM54654443
Additional Details : Weight : 1.57000kg
 View more versions of this product
Product Code. 15958741
  • 128.26 EUR / 1 pair
Estimated Shipment: 27-09-2024
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • Lace-up front with zipper covering
  • GripForce™ Easy Twist Outsole
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
43
Laces
Vila
Microfiber
Steel