Learn More
AKOS B018304, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
AKOS B018304 is an arylalkylidene derivative, with polar substitution at para-position.
Specifications
Specifications
| Chemical Name or Material | AKOS B018304 |
| Physical Form | Solid |
| CAS | 6308-22-1 |
| Color | Gray |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H6N2O3S2 |
| Solubility Information | DMSO : ≥ 77.5 mg/mL (291.03 mM) |
| SMILES | O=C1NC(S/C1=C/C2=CC=CC=C2[N+]([O-])=O)=S |
| Molecular Weight (g/mol) | 266.3 |
| Show More |