Learn More
Heidolph™ Vial PolyBlock
Accommodate specifically sized vessels for your StarFish magnetic stirrer workstation. Heidolph™ Vial PolyBlock, is one of up to 5 StarFish PolyBlock segments that can be mixed at a single stirrer workstation to create various arrays of vessel size and type.
Brand: Heidolph™ 5058002800
Additional Details : Weight : 0.01000kg
Description
- Accommodates one specific vial size.
- One of up to 5 segments per StarFish array.
Specifications
PolyBlock | |
Starfish workstation, MR-Hei magnetic stirrers | |
9 x 15 mm Vials |