missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph instruments Vial MonoBlock
Accommodate specifically sized reagent tubes at your StarFish magnetic stirrer workstation. Heidolph™ Vial MonoBlock, constructed of one piece, is ideal for experiments using the same vessel.
515.00 EUR
Specifications
Description | MonoBlock |
---|---|
For Use With (Equipment) | Starfish Workstation, MR-Hei Magnetic Stirrers |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11771699
|
Heidolph instruments
505-80006-00 |
16 x 28 mm Vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781699
|
Heidolph instruments
505-80007-00 |
20 x 21 mm vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11791699
|
Heidolph instruments
505-80008-00 |
40 x 17 mm Vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11701709
|
Heidolph instruments
505-80009-00 |
4 x 15 mm Vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11711709
|
Heidolph instruments
505-80010-00 |
40 x 12 mm Vials |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Accommodates one specific vial size.
- One-piece construction.
Specifications
MonoBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers |