Learn More
Saint-Gobain Versilic™ Silicone Stoppers, 35mmH
Brand: Saint-Gobain 26303805
Additional Details : Weight : 0.14000kg
Description
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
Cap | |
31 mm | |
5/Pk. | |
–60 to 200°C |
35mm | |
38 mm | |
Silicone |