Learn More
Saint-Gobain Versilic™ Silicone Stoppers, 35mmH
Brand:  Saint-Gobain 26304105
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11744578
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
Specifications
Specifications
Silicone