missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilic™ Silicone Sealing Cap
Offers a very secure hold and is comfortable to use. Versilic™ Silicone Sealing Cap has a slight taper to maximize seal.
Brand: Saint-Gobain 162016-50
Additional Details : Weight : 0.80000kg
Description
- Versilic™ Silicone construction
- Transparent
- Taperd to maximize seal
- For use with test tubes
Specifications
Test Tube | |
16 mm | |
50/Pk. |
Versilic Cap | |
Transparent | |
-60°C to +220°C |