Learn More
Ansell™ VersaTouch™ 92-220
12.66 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
7
8
9
10
Description
- Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions.
- A thinner glove with enhanced fingertip flexibility and improved overall comfort.
- Textured fingertips provide good comfort and tactility for sensitive, precise handling.
- Latex-free: no risk of Type 1 allergies.
- Approved for food contact applications. Ideal for handling all fatty food.
Specifications
Exterior Finish | Fully Textured |
Material | Nitrile |
Size | 10 |
Thickness (English) | 2.9 mil |
Thickness (Metric) | 0.07 mm |
Model | 9222010 |
Series | 92-220 |