Learn More
Ansell™ VersaTouch™ 37-501 Series Blue Nitrile Immersion Gloves
57.62 EUR valid until 2024-12-31
Use promo code "21907" to get your promotional price.
Use promo code "21907" to get your promotional price.
Click to view available options
Size:
6.5
7.5
8.5
9.5
10.5
Description
- Excellent resistance to many solvents and oil based chemicals
- Very good abrasion resistance
- Flexible and comfortable for an excellent dexterity
- Gauntlet cuff style
- Raised diamond grip
- Powder free
Specifications
Back Material | Nitrile |
Cuff Material | Nitrile |
Product Type | Immersion Gloves |
Size | 10.5 |
Style | 37-501 |
Thickness (Metric) | 0.42 mm |
Glove Cuff Color | Blue |
Glove Cuff Style | Gauntlet |
Liner Material | Cotton Flocking |
Quantity | 12 Pairs |