Learn More
Sartorius Stedim Biotech™ Vacuum Hose With Quick Con., Silicone
Flexible and durable quick connecting silicone hose for Combisart vacuum manifold system.
Brand: Sartorius Stedim Biotech™ 1ZAS--0016
50.07 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- For use with Combisart vacuum manifold system
- Durable
- Flexible
- Resistant
- Quick connect
Specifications
Silicone | |
1 m | |
Combisart vacuum manifold system | |
Quick connect |