Learn More
Wheaton™ Vaccum Filter with Frit
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Brand:  Wheaton™ 419452
726.75 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11751883
  • 765.00 EUR / Each
Estimated Shipment: 22-05-2024
Description
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Specifications
Specifications
Large Sample Volumes, Liquids With A Heavy Particulate Load And Viscous Liquids When A High Flow Rate Is Required
Vaccum Filter
90 mm