Learn More
Wheaton™ Vaccum Filter with Frit
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Brand:  Wheaton™ 419520-47
698.25 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.36080kg
 View more versions of this product
Product Code. 10498711
  • 735.00 EUR / Each
Estimated Shipment: 23-05-2024
Description
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Specifications
Specifications
47 mm
To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Vaccum Filter
47 mm