Learn More
Fisherbrand™ UV Lamp, Dual Wavelength
Used as a method of purification. Fisher Scientific™ UV Lamp, Dual Wavelength can be used in combination with other methods to reduce microbial reproduction in purified water.
352.00 EUR - 354.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15449689
|
Fisherbrand™
|
Accu100 Ultrapure Water System |
354.00 EUR
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
15469689
|
Fisherbrand™
|
Accu500 Ultrapure Water System |
352.00 EUR
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
Description
- For use with Accu100 and Accu500 Ultrapure Water Systems
- Prevents microbial reproduction
Specifications
185/254nm |