missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine-5'-diphosphoglucose, disodium salt, 95%, MP Biomedicals™
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Specifications
Color | White |
Assay Percent Range | 95% |
Quantity | 100 mg |
Formula Weight | 610.3 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Uridine-5'-diphosphoglucose, disodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'