Learn More
Uric acid, +99%, Thermo Scientific Chemicals
Catalog Number p-210731
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
69-93-2
C5H4N4O3
168.112
MFCD00005712
LEHOTFFKMJEONL-UHFFFAOYSA-N
uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro
1175
CHEBI:17775
7,9-dihydro-3H-purine-2,6,8-trione
C12=C(NC(=O)N1)NC(=O)NC2=O
69-93-2
C5H4N4O3
168.112
MFCD00005712
LEHOTFFKMJEONL-UHFFFAOYSA-N
uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro
1175
CHEBI:17775
7,9-dihydro-3H-purine-2,6,8-trione
C12=C(NC(=O)N1)NC(=O)NC2=O
Uric acid, CAS # 69-93-2, is a common oxopurine metabolite, and a normal component of urine.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Uric acid is a purine-based metabolite found in humans, mice and E. coli
• Uric acid can accumulate in excess leading to diseases such as hyperuricemia
• Effects of excess uric acid accumulation include hypertension, arthritis, and renal issues
Applications
• Useful as a biochemical standard
• Suitable for use in the study of hyperuricemia and gout
• Uric acid may also play other roles in cell metabolism and physiology
• Intended for research use only
TRUSTED_SUSTAINABILITY
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 25 g
Formula Weight 168.11
Percent Purity ≥99.0% (ex N)
Physical Form Fine Powder
Chemical Name or Material Uric acid, 99+%