missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog Number p-3737727
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
57-13-6
CH4N2O
60.06
MFCD00008022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
57-13-6
CH4N2O
60.06
MFCD00008022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance (8M in H2O), (8 M in H2O) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm
Melting Point 131.0°C to 135.0°C
Color White
Assay Percent Range 99.5% min
Packaging Glass Bottle
Linear Formula H2NCONH2
Quantity 1 kg
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol, benzene, concentrated hcl, practically insoluble in chloroform and ether, soluble in methanol: 16.6 g/100 mL, soluble in ethanol: 5 g/100 mL
Formula Weight 60.06
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Solid
Chemical Name or Material Urea, DNAse, RNAse and Protease Free
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'