Learn More
Cole-Parmer™ Ultrapure Application Filter Cartridges
Tolerant to an accumulated exposure of 10 hours. Filters may also be sanitized with compatible chemical agents.
Brand:  Cole-Parmer™ 06479-52
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11726119
  • 555.00 EUR / Each
Estimated Shipment: 15-07-2024
Description
  • These cartridges are autoclavable to 250°F 121°C) and in-line sterilizable to 257°F (125°C) for up to 30-minute cycles
  • Tolerant to an accumulated exposure of 10 hours
  • Filters may also be sanitized with compatible chemical agents
  • Suitable to 180°F (82°C) at 10psid
  • The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121°C Plastics guidelines
  • Pass the MEM Elution Cytotoxicity Test
  • Aqueous extracts contain less than 0.25 EU/mL
  • Made of resin-bonded borosilicate glass microfibers that do not leach flavor-altering substances
  • Meet NSF Standard 53 for the reduction of cysts
  • Ideal as prefilters to protect membrane filter/RO systems
Specifications
Specifications
Double open end PTFE filter cartridge, 0.2µm
PTFE
Ultrapure Applications
10 in.
PTFE
5.9 ft.2
Ultrapure Application Filter Cartridge
25.4 cm