Learn More
Masterflex™ Tygon SE–200 Tubing
Brand: Masterflex™ 95713-30
Additional Details : Weight : 2.00000kg
Specifications
FDA Compliant for contact food products | |
Tygon SE-200 | |
15.24 m | |
Chemical and pharmaceutical processing, beverage dispensing, paint and solvent production, semiconductor processing | |
1 Each | |
Transparent | |
74°C |
9.6 mm | |
9.6 mm | |
2.3 mm | |
-45°C | |
Sterile | |
50 psi |