missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human TXNL1.
Brand: Abnova PAB29376
Additional Details : Weight : 0.02000kg
Description
Sequence: CLNESDEHGFDNCLRKDTTFLESDCDEQLLITVAFNQPVKLYSMKFQGPDNGQGPKYVKIFINLPRSMDFEEAERSEPTQALELTEDDIKEDGIVPLRYVKFQNVNSVTIFVQSNQGEEETTRISYFTFIGTPVQATNMNDFKRVVGKSpecifications
TXNL1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human TXNL1. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
TXNL1 | |
Recombinant protein corresponding to human TXNL1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
TRP32/TXL-1/TXNL/Txl | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9352 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |