Learn More
WTW™ Turbidity Standard
Turbidity standards AMCO Clear™ with the highest precision and long-term stability are N.I.S.T. traceable, lot certified and not hazardous to your health.
450.00 EUR - 570.00 EUR
Specifications
Type | Turbidity Standard |
---|---|
For Use With (Equipment) | Turb 555 IR |
Description | Calibration standards |
Product Code | Brand | Turbidity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Turbidity | Price | Quantity & Availability | |||||
10478413
|
WTW™
600543 |
0.02, 10, 100,1750 NTU |
570.00 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10134273
|
WTW™
600544 |
10,000 NTU |
450.00 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
- Long-term precision and stability
- No health hazards
- Easy to dispose of
- Adapted to the respective device optics in their particle composition
- Suitable for applications in the lower measuring range such as drinking water
Specifications
Turbidity Standard | |
Turb 555 IR | |
Calibration standards |