missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebro Electronic™ TTX 110 Thermometer
Thermometer, type T with probe
Brand: Ebro Electronic™ 1340-5110A
Additional Details : Weight : 0.15930kg
Description
Therm. pointed probe (Needle length: 120 mm, dia. 3 mm), cable 0.6mSpecifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16 | |
Single-use | |
17 mm | |
200 mm | |
Stainless Steel and Plastic | |
Permanently attached pointed | |
0.18 kg | |
None | |
350°C | |
-50°C | |
Factory Calibrated | |
Lithium CR 2032 | |
90 mm | |
100 mm |
3 mm | |
Thermometer to measure ambient or heart | |
Thermometer, sensor, calibration certificate, instruction manual | |
105 mm | |
TTX 110 | |
0.1°C from −50 to 199.9°C, 1°C above | |
Core Thermocouple Thermometer | |
662°F | |
-58°F | |
-50°C to +350°C | |
Typically 100 hours of uninterruped use | |
±0.8°C | |
LCD |