Learn More
TSTD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554601
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MPSSTSPDQG DDLENCILRF SDLDLKDMSL INPSSSLKAE LDGSTKKKYS FAKKKAFALF VKTKEVPTKR SFECKEKLWK CCQQLFTDQT SIHRHVATQH ADEIYHQT Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
TSTD2 is a protein coding gene.Specifications
TSTD2 | |
Polyclonal | |
Unconjugated | |
TSTD2 | |
C9orf97; PP4189; rhodanese domain-containing protein 2; rhodanese like domain containing 2; thiosulfate sulfurtransferase (rhodanese)-like domain containing 2; thiosulfate sulfurtransferase like domain containing 2; thiosulfate sulfurtransferase -like domain containing 2; thiosulfate sulfurtransferase/rhodanese-like domain-containing protein 2; TSTD2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158427 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T7W7 | |
TSTD2 | |
Recombinant protein corresponding to Human TSTD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only