missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin I Type 3 (cardiac) Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Bio-Techne NBP3-11029F
This item is not returnable.
View return policy
Description
Troponin I Type 3 (cardiac) Polyclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for ELISA
Specifications
Troponin I Type 3 (cardiac) | |
Polyclonal | |
ELISA | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
Purified Human Cardiac Troponin, I subunit | |
0.1 mL | |
Apoptosis, Cell Biology, Cellular Markers, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
7137 | |
Store at 4°C in the dark. |
ELISA | |
FITC | |
PBS | |
Goat | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction