missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Assay Percent Range | >99.9% |
Formula Weight | 121.1 |
Grade | Ultrapure |
Chemical Name or Material | Tris |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'