Learn More
Triphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Catalog Number p-208866
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
6399-81-1
C18H16BrP
343.20
MFCD00035107
CMSYDJVRTHCWFP-UHFFFAOYSA-N
triphenylphosphine hydrobromide, triphenylphosphonium bromide, phosphine, triphenyl-, hydrobromide, triphenylphosphane hydrobromide, triphenylphosphine bromine, pubchem13868, ph3p hbr, ph3p . hbr, unappfzuzwmopv-uhfffaoysa-n
80811
triphenylphosphane;hydrobromide
Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
6399-81-1
C18H16BrP
343.20
MFCD00035107
CMSYDJVRTHCWFP-UHFFFAOYSA-N
triphenylphosphine hydrobromide, triphenylphosphonium bromide, phosphine, triphenyl-, hydrobromide, triphenylphosphane hydrobromide, triphenylphosphine bromine, pubchem13868, ph3p hbr, ph3p . hbr, unappfzuzwmopv-uhfffaoysa-n
80811
triphenylphosphane;hydrobromide
Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Melting Point 196.0°C to 199.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Fieser 15,353
Formula Weight 343.2
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Triphenylphosphonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Triphenylphosphonium bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.