Learn More
Triethyl phosphite, 98%, Thermo Scientific Chemicals
Catalog Number 10676394
Click to view available options
Quantity:
100 mL
500 mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
122-52-1
166.16
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
ethyl phosphite, eto 3p, hsdb 895, triethoxyphosphine, unii-6b2r04s55g, c2h5o 3p, p oet 3, phosphorous acid, triethyl ester, fosforyn trojetylowy czech, triethylphosphite, phosphorous acid triethyl ester
31215
triethyl phosphite
CCOP(OCC)OCC
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
122-52-1
166.16
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
ethyl phosphite, eto 3p, hsdb 895, triethoxyphosphine, unii-6b2r04s55g, c2h5o 3p, p oet 3, phosphorous acid, triethyl ester, fosforyn trojetylowy czech, triethylphosphite, phosphorous acid triethyl ester
31215
triethyl phosphite
CCOP(OCC)OCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -112.0°C
Density 0.9650 g/mL
Boiling Point 156.0°C to 158.0°C
Flash Point 44°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15O3P
Linear Formula (C2H5O)3P
Refractive Index 1.4120 to 1.4140
Quantity 500 mL
Beilstein 01, 330
Fieser 01,1212; 02,432; 03,304; 04,529; 05,693; 06,612; 07,387; 08,501; 11,555; 12,143
Merck Index 15, 9840
Specific Gravity 0.965
Solubility Information Solubility aq. soln.: slightly soluble (hydrolysis). Other solubilities: soluble in alcohol and ether
Viscosity 1 mPa.s (20°C)
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Triethyl phosphite, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl phosphite
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.