Learn More
enQuireBio™ TranslationBlocker Human TLR7 / CD287 siRNA
Blocks transcription and translation of TLR7 / CD287 from Human.
Brand:  enQuireBio™ QX26-10nmol
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16089191
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_057646
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
51284
10 nmol
Human
293T Human Embryonic Kidney Cells with Large T Antigen.
TLR7
siRNA
TLR7-like, TLR7, Toll-like receptor 7, TLR-7
Lyophilized