Learn More
enQuireBio™ TranslationBlocker Human Interleukin-10 / IL-10 siRNA
Blocks transcription and translation of Interleukin-10 / IL-10 from Human.
Brand: enQuireBio™ QX36-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_000563 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3586 | |
2 nmol | |
Human |
Dendritic Cells derived from adherent human PBMCs. | |
IL10 | |
siRNA | |
IL10A, IL-10, TGIF, CSIF, GVHDS | |
Lyophilized |