missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-4-(Boc-amino)cyclohexanecarboxylic acid, 98%

Brand:  Alfa Aesar™ H59402.01

Additional Details : CAS Number : 53292-89-0 Weight : 0.00025kg

Quantity 0.25g

 View more versions of this product

Product Code. 15445006

  • 93.00 EUR / 0.25g


Estimated Shipment date
from Supplier 24-06-2019
Add to basket

Specifications

Chemical Identifiers

53292-89-0
243.303
KXMRDHPZQHAXML-UHFFFAOYSA-N
2755996
CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O
C12H21NO4
MFCD01862293
trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid

Specifications

trans-4-(Boc-amino)cyclohexanecarboxylic acid
0.25g
C12H21NO4
7641071
KXMRDHPZQHAXML-UHFFFAOYSA-N
4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
2755996
Powder
White
53292-89-0
MFCD01862293
trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O
243.303
243.3
98%
Videos
SDS Documents