missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-3,5-Difluorocinnamic acid, 99%

Brand:  Alfa Aesar™ H61868.06

Additional Details : CAS Number : 147700-58-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458458

  • 146.00 EUR / 5g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

147700-58-1
184.142
MBAWRXICVNIUGY-OWOJBTEDSA-N
5374941
C1=C(C=C(C=C1F)F)C=CC(=O)O
C9H6F2O2
MFCD00010321
3,5-difluorocinnamic acid, trans-3,5-difluorocinnamic acid, 3,5-difluorocinnamicacid, e-3-3,5-difluorophenyl acrylic acid, 3-3,5-difluorophenyl acrylic acid, 2e-3-3,5-difluorophenyl prop-2-enoic acid, 2e-3-3,5-difluorophenyl acrylic acid, rarechem bk hw 0255, timtec-bb sbb006675, 3-3,5-difluorophenyl propenoic acid
(E)-3-(3,5-difluorophenyl)prop-2-enoic acid

Specifications

trans-3,5-Difluorocinnamic acid
204°C to 205°C
147700-58-1
MFCD00010321
MBAWRXICVNIUGY-OWOJBTEDSA-N
(E)-3-(3,5-difluorophenyl)prop-2-enoic acid
5374941
Powder
White
5g
C9H6F2O2
3,5-difluorocinnamic acid, trans-3,5-difluorocinnamic acid, 3,5-difluorocinnamicacid, e-3-3,5-difluorophenyl acrylic acid, 3-3,5-difluorophenyl acrylic acid, 2e-3-3,5-difluorophenyl prop-2-enoic acid, 2e-3-3,5-difluorophenyl acrylic acid, rarechem bk hw 0255, timtec-bb sbb006675, 3-3,5-difluorophenyl propenoic acid
C1=C(C=C(C=C1F)F)C=CC(=O)O
184.142
184.14
99%
Videos
SDS