Learn More
Fisherbrand™ Traceable™ AC Adapter
Traceable™ AC Adapter simplifies the use of thermometers, and hygrometers
Brand: Fisherbrand™
Additional Details : Weight : 0.18140kg
Description
- Increase the ease of use on Traceable™ products
- Avoid costly battery changes with a universal charger
Specifications
Traceable Scientific Thermometers | |
Cord and plug set | |
0.18 kg | |
Traceable Scientific Thermometers 90080-09, 37804-06, and 37804-07 | |
100/240 V |
4.572 cm | |
8.4 cm | |
Universal/International AC Adaptor | |
6.858 cm | |
50/60 Hz |