Learn More
Topiramate, 98%, Thermo Scientific Chemicals
Catalog Number p-7077606
Click to view available options
Quantity:
1 g
5 g
97240-79-4
C12H21NO8S
339.36
KJADKKWYZYXHBB-XBWDGYHZSA-N
topiramate, topamax, epitomax, topamax sprinkle, tipiramate, topiramatum, tipiramato, topiramato, tipiramate french, tipiramato spanish
5284627
CHEBI:63631
[(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
CC1(OC2COC3(C(C2O1)OC(O3)(C)C)COS(=O)(=O)N)C
97240-79-4
C12H21NO8S
339.36
KJADKKWYZYXHBB-XBWDGYHZSA-N
topiramate, topamax, epitomax, topamax sprinkle, tipiramate, topiramatum, tipiramato, topiramato, tipiramate french, tipiramato spanish
5284627
CHEBI:63631
[(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
CC1(OC2COC3(C(C2O1)OC(O3)(C)C)COS(=O)(=O)N)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 125.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 339.36
Percent Purity 98%
Chemical Name or Material Topiramate
compliance-icons
Product Identifier
  • Topiramate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.