Learn More
RETSCH Titanium Push-fit Rotor
Ensure the gentle preparation of analytical samples in a very short time. Retsch™ Titanium Push-fit Rotors can be used for a variety applications.
Brand:  RETSCH 02.608.0047
Additional Details : Weight : 0.30000kg
Product Code. 10100791
  • 3075.00 EUR / Each
Estimated Shipment: 09-07-2024
Description
  • 12-tooth rotor
  • For use with ZM 200 Model Ultra-Centrifugal Mill
Specifications
Specifications
Push-fit Rotor
For Heavy-metal-free Grinding
For Heavy-metal-free Grinding
Titanium