Learn More
Lovibond™ Tintometer™ VARIO™ Phosphonates Reagent Set, 0 to 125mg/L
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
Brand: Lovibond™ 535220
Additional Details : Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
- Range: 0 to 125mg/L
- Parameter: Phosphonates
- Method: UV Oxidation
- For use in our test kits, colorimeters and spectrophotometers
- Case of 100
Specifications
Water Testing | |
Case of 100 | |
UV Oxidation |
Test kits, colorimeters and spectrophotometers | |
0 to 125mg/L | |
Phosphonates Reagent Set |