Learn More
Lovibond™ Tintometer™ VARIO™ Phosphonates Reagent Set, 0 to 125mg/L
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
Brand:  Lovibond™ 535220
Additional Details : Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 15361685
  • 133.00 EUR / Each
Estimated Shipment: 19-08-2024
Description
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
  • Range: 0 to 125mg/L
  • Parameter: Phosphonates
  • Method: UV Oxidation
  • For use in our test kits, colorimeters and spectrophotometers
  • Case of 100
Specifications
Specifications
Water Testing
Case of 100
UV Oxidation
Test kits, colorimeters and spectrophotometers
0 to 125mg/L
Phosphonates Reagent Set