missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Nitrite No. 1 Tablets, foil pack
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
36.80 EUR - 44.70 EUR
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Quantity | 100 Tablets |
Type | Nitrite No. 1 Tablets |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15371815
|
Lovibond™
515220BT |
100 Tablets |
36.80 EUR
Pack of 250 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15351815
|
Lovibond™
515200BT |
100 Tablets |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Nitrite No. 1 Tablets |