Learn More
Lovibond™ Tintometer™ Hardness LR Test
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505171
Additional Details : Weight : 0.30000kg Transport : UN number : 3259 Chem class : 8 Pack group : III
Description
- Hardness LR Test (BW)
- Bottle of 250
Specifications
Water Testing | |
Test kits, colorimeters and spectrophotometers | |
250 Tablets | |
Hardness LR Test (BW) |
Safety and Handling
- X250 HARDNESS LR (BW) TABLETS
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : cellulose, 2-amino-2-methylpropane-1,3-diol, trisodium phosphsate dodecahydrate