missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Description
Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Odor | Alcohol-like |
Linear Formula | Sn[OCH(CH3)2]4 |
Quantity | 25 mL |
UN Number | UN3274 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with alcohols,ketones and esters. |
Formula Weight | 355.06 |
Percent Purity | 99% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'