Learn More
Cole-Parmer™ Threaded Reducing Nipples
Threaded fittings offer more permanent installation
Brand: Cole-Parmer™ 06546-23
Additional Details : Weight : 0.80000kg
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Threaded Male Reducer | |
Autoclavable | |
1/8 in. | |
Autoclave or Ethylene Oxide | |
Translucent |
NPT(M) Connection | |
PFA | |
1/4 in. | |
1/4 in. x 1/8 in. NPT(M) |