Learn More
Masterflex™ Thread to Compression Adapters
Compression fittings are an excellent choice for connecting rigid tubing systems
73.85 EUR - 120.00 EUR
Specifications
Product Type | Thread to Compression Adapter |
---|---|
Description | Compression x NPT(M) Connection |
Autoclavable | Not Autoclavable |
Material | Polypropylene |
Packaging Quantity | 10 Fittings |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
11791973
|
Masterflex™
06385-78 |
NPT(M) 3/4 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781973
|
Masterflex™
06385-76 |
NPT(M) 1/2 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Available in packs of ten
Specifications
Thread to Compression Adapter | |
Not Autoclavable | |
10 Fittings | |
3/4 in. | |
Straight |
Compression x NPT(M) Connection | |
Polypropylene | |
3/4 in. | |
Ethylene Oxide |