Learn More
Thin Insulating Tubes, Thermo Scientific™
Oxide ceramics tubes resistant to corrosive liquids and gases
Brand:  Thermo Scientific Alfa Aesar 031489.DM
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11373887
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Inert to oxidation
  • Not subject to radiation damage
  • Wear resistant and hard
  • With both ends open
Specifications
Specifications
100 mm
1.5mm
2mm