Learn More
Thimerosal, Fisher BioReagents
Catalog Number p-28875
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
54-64-8
C9H9HgNaO2S
404.81
MFCD00013062
RTKIYNMVFMVABJ-UHFFFAOYSA-L
Sodium ethylmercurithiosalicylate
16684434
CHEBI:9546
sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
[Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
54-64-8
C9H9HgNaO2S
404.81
MFCD00013062
RTKIYNMVFMVABJ-UHFFFAOYSA-L
Sodium ethylmercurithiosalicylate
16684434
CHEBI:9546
sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
[Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O
  • Thimerosal is a mercury-containing compound commonly used as an antibacterial and antifungal agent
  • Also used as a bactericide in pharmaceuticals and research
  • Intended for research use only
TRUSTED_SUSTAINABILITY
Type Thimerosal
Physical Form Solid
pH 6.7 to 8.2
Melting Point 232°C
Assay Percent Range ≥97 %
Loss on Drying 0.5% max.
Merck Index 15, 9470
Packaging Amber Glass
Formula Weight 404.8
Percent Purity 97% min.
Color Cream
Quantity 10 g
Chemical Name or Material Thimerosal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thimerosal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.