Learn More
Ebro Electronic™ Thermocouple Surface Probes
General purpose surface probes, Surface temperature measurement of motors, turbines, pumps, casting, molds, heating tubes, heating plates, injection molding, heating boilers, incinerators etc.
457.00 EUR - 740.00 EUR
Specifications
Certifications/Compliance | RoHS Directive 2011/65/EU |
---|---|
Disposable | Single-use |
Length (Metric) | 300 mm |
Material | Metal |
For Use With (Equipment) | TFN 520 |
Product Code | Brand | Probe Type | Length (Metric) Probe | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Probe Type | Length (Metric) Probe | For Use With (Application) | Price | Quantity & Availability | |||
11753489
|
Ebro Electronic™
1341-0639 |
Roller Moving | 66 mm | Rail surface probes. Temperature measurement of moving surfaces. |
740.00 EUR
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
10636711
|
Ebro Electronic™
1341-0710 |
Thermocouple Type K | 30 mm | General purpose surface probes. Surface temperature measurement of motors, turbines, pumps, casting molds, heating tubes, heating plates, injection molding, heating boilers, incinerators etc. |
457.00 EUR
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
Specifications
RoHS Directive 2011/65/EU | |
300 mm | |
TFN 520 |
Single-use | |
Metal |