Learn More
Ebro Electronic™ Thermocouple Penetration Probes
Penetration probes for temperature measurement of viscoplastic masses such as asphalt, bitumen or grounds.
Brand: Ebro Electronic™ 1341-0674
Additional Details : Weight : 0.80000kg
Specifications
RoHS Directive 2011/65/EU | |
Single-use | |
30 mm | |
1 m | |
Penetration | |
Penetration Probe | |
500°C | |
TFN 520 |
2.1 mm | |
Penetration probe, temperature measurement of viscoplastic masses such as asphalt, bitumen | |
250 mm | |
TPN 230 | |
0.12 kg | |
500°C | |
-200°C to 500°C | |
100 mm |