Learn More
Invitrogen™ Mouse/Rat/Guinea Pig LAMP2 (aa 400-411) synthetic peptide Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP039
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Rat, Mouse, Guinea Pig LAMP-2 (residues 400-411) Synthetic Peptide, PEP-039, from Invitrogen.
PEP-039 is a 13 amino acid synthetic peptide whose sequence corresponds to cytoplasmic domain amino acid residues 400-411 of rat LAMP-2 protein. This sequence is completely conserved between rat, mouse, and guinea pig LAMP-2. The sequence of this peptide is (amino to carboxy terminus): (C)-G400-L-K-R-H-H-T-G-Y-E-Q-F411 (A cysteine residue was added to facilitate coupling KLH).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and rat LAMP-2, catalog # PA1-655. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
100717075, 16784, 24944 | |
purified | |
Mouse/Rat/Guinea Pig LAMP2 (aa 400-411) synthetic peptide | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Guinea Pig, Mouse, Rat | |
P17046, P17047 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Lamp2 | |
No preservative | |
50 μg | |
(C) - G400 - L - K - R - H - H - T - G - Y - E - Q - F411 |