missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microplate Readers Optical Filters

Brand:  Thermo Scientific™ 1425600

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11749428

  • 197.00 EUR / Each


Estimated Shipment
01-02-2021
Add to basket

Description

Description

  • All filters have a 3nm bandpass centered at the specified wavelength
  • Compatible with all three Multiskan models

Specifications

Specifications

560nm
Videos
SDS
Documents