Learn More
Thermo Scientific™ Clear Seal Heat Sealing Film
Ideal for applications requiring pierceable non-conductive seal material
Brand: Thermo Scientific™ AB-1797
728.89 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.
Additional Details : Weight : 1.20600kg
Description
Ideal for applications requiring pierceable non-conductive seal material
- Suitable for PCR applications with screw- or clip-down heated lid thermal cyclers only
- Seals polypropylene, polystyrene, polyethylene and Diamond plates
- Easily pierced and peelable by hand from 96-well plates
- Temperature range: -80°C to +80°C
Specifications
Clear Seal Thin, Polymer 85 x 135 | |
100 Sheets |