Learn More
Thermo Scientific™ ART™ Non-Filtered Pipette Tips
Range of styles and packaging options
Brand: Thermo Scientific™ 3900
Additional Details : Weight : 0.29250kg
Description
Range of styles and packaging options
- All sterile HLT branded tips are certified “Pure”
- All nonsterile tips are manufactured in clean room environments and are certified DNA, RNase and DNase free
- Stringent manufacturing and QA/QC testing procedures assure the most aseptic products on the market
- Some tips have reference marks which provide a quick and convenient guide to visually establish whether you are pipeting the desired volume of liquid
Specifications
200 μL | |
Non-sterile | |
Bulk | |
ART |
Non-Filtered | |
Yellow | |
Standard |