Learn More
Brady™ Thermal Transfer Printable Flags, 76 mm Core
Flags for Thermal Transfer Printers.
Brand:  Brady™ 217024
Additional Details : Weight : 0.22400kg
 View more versions of this product
Product Code. 11588624
  • 149.00 EUR / 1 roll
Estimated Shipment: 31-05-2024
Description
  • Excellent solvent resistance
  • For smooth surfaces
  • Permanent adhesive
Specifications
Specifications
Not Autoclavable
White
Polypropylene (B-425A)
Non-sterile
BBP™11, BBP™12, BradyPrinter PR Plus Series
1000 Labels
-70°C to +80°C
26 mm
30 mm
10 mm
Acrylic
Matte
Thermal Transfer
Asset Tracking, Barcode Labeling, Equipment Labeling, General Identification, Inventory Labeling, Laboratory Labeling
1
Rectangle
0.11 mm
30 mm
10 mm