Learn More
Ebro Electronic™ TFX 430 Precision Thermometer
Reference thermometer with exchangeble PT 100 probe
Brand: Ebro Electronic™ 1340-5438A
Additional Details : Weight : 0.80000kg
Description
TFX 430 Model Precision Thermometer. Powered by a replaceable lithium battery, these precision thermometers are handy, watertight temperature measurement devices for a whole range of measurement and control applications in the laboratory and in industry.Specifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
Single-use | |
54 mm | |
355 mm | |
ABS Plastic | |
Pointed | |
Precision Thermometer | |
932°F | |
-148°F | |
-100°C to +500°C | |
AG 130, AG 140, AG 150, AX 110, TPX 130, TPX 230, TPX 330 | |
Lithium 3 V/Typ CR 2477 | |
120 mm | |
162 mm |
3 mm | |
Applications: Laboratory, temperature monitoring, measure air, fluids, ductile substances, aggressive media and surfaces, Process monitoring | |
Thermometer, sensor, calibration certificate, instruction manual | |
130 mm | |
TFX 430 and TPX 230 | |
90 g | |
None | |
500°C | |
-100°C | |
Factory Calibrated | |
Approx. 5 Years | |
±0.05°C/±0.09°F | |
LCD |