Learn More
Ebro Electronic™ TFX Core Thermometer Series
Precise and very robust thermometer with a big display
Brand: Ebro Electronic™ 1340-5418A
Additional Details : Weight : 0.34200kg
Description
Thermometer for various applications in food, laboratory, industrial use. All thermometers includes a factory calibration certificate. The thermometer measures with PT 1000 temperature sensor. Core thermometer, precision, lithium battery, watertight, measurement, temperature, laboratory, sensor, display, robust, impact resistant, accuracy.Specifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16; EN 13485 | |
For bakeries, butcheries and for the food industry | |
Thermometer, sensor, calibration certificate, instruction manual | |
ABS Plastic | |
Blunt | |
90 g | |
None | |
572°F | |
-58°F | |
-50°C to +300°C or -50°C to +400°C | |
AG 130, AG 140, AG 150, AG 160, AX 100 | |
Lithium 3 V/Typ CR 2477 | |
120 mm | |
50 mm |
Single-use | |
52 mm | |
335 mm | |
TFX 410-1 & TPX 200 | |
0.1°C | |
Precision Core Thermometer | |
Without cable | |
300°C | |
-50°C | |
Factory Calibrated | |
Approx. 5 Years | |
±0.3°C | |
LCD |